Precoiled Pipe Markers 'Heating Water Supply', Black/Yellow, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A4056 B4056 C4056 D4056 E4056 F4056 G4056 H4056
94003-392EA 28.11 CAD
94003-392 94004-166 94005-592 94009-520 94012-654 94013-850 94014-396 94015-176
Precoiled Pipe Markers 'Heating Water Supply', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007