Dissolution Baskets and Basket Accessories for Apparatus 1
Supplier: QLA
BSK010-01 BSK010-HR BSK020-01 BSK020-DK BSK040-01 BSK040-DK BSKHLD-12 BSKPTD-40 BSKPTH-40 BSKPTV-40 BSKSUP-01 BSKSUP-DK BSKSUP-HR BSK040-ZM BSK040-ZM2 BSK100-DK BSK010-CA BSK010-DKC BSK020-CA BSKSUP-EW BSK100-01 BSK010-EW BSK040-HRP BSK100-HR BSK010-DK BSK010-PT BSK010-ST BSK020-EW BSK020-PT BSK020-ST BSK040-CA BSK040-EW BSK040-PT BSK040-ST BSKSUP-CA BSKSUP-PT BSKSUP-ST BSK040-DKC BASKETWASH BSKPTP-40 BSKTOOL BSK080-ST BSK100-DKC BSK100-ST BSK140-CA BSK140-HR BSK140-ST BSK150-01 BSK150-EW BSKHLD-6 CUSBSK20150DKC CUSBSK-20150EW CUSBSK-20150HR CUSBSK-20150PT CUSBSK-20100DK CUSBSK20100DKC CUSBSK-20100EW CUSBSK-20100HR CUSBSK-20100PT CUSBSK-20100ST CUSBSK-20100VK CUSBSK-20150DK CUSBSK-20150ST CUSBSK-20150VK BSK030-01 BSK030-DKC BSK060-DKC BSK080-DKC
11010-012EA 514.35 CAD
11010-012 11010-014 11010-016 11010-018 11010-022 11010-024 11010-028 11010-030 11010-032 11010-034 11010-044 11010-046 11010-048 89039-780 89049-186 89049-188 89080-274 89080-276 89080-278 89080-282 89106-788 89132-956 89132-958 89173-102 93001-046 93001-048 93001-050 93001-052 93001-054 93001-056 93001-058 93001-060 93001-062 93001-064 93001-066 93001-068 93001-070 97002-352 75848-828 75848-830 75848-844 75848-998 75849-000 75849-002 75849-004 75849-006 75849-008 75849-010 75849-012 75849-014 75849-042 75849-044 75849-046 75849-048 75849-162 75849-164 75849-166 75849-168 75849-170 75849-172 75849-174 75849-176 75849-178 75849-180 75849-242 75849-244 75849-246 75849-248
Dissolution Baskets and Basket Accessories for Apparatus 1
Dissolution Apparatus
QLA is the only on-site basket manufacturer in the industry.
  • Made of high quality 316 Stainless Steel (excludes Suppository Baskets)
  • All baskets made in the USA
  • Laser marked with unique serial number (excludes Suppository baskets)
  • 40 Mesh baskets are made from sintered mesh
  • Variety of mesh sizes available
QLA offers both clip style and O-ring Push-on style baskets for most dissolution baths.
Our 40 mesh baskets are made with sintered 316 Stainless Steel. The sintering process essentially creates a welded joint at each overlapping wire junction.More Product Information