XPert™ Bulk Powder Filtered Stations, Labconco®
Supplier: Labconco
3962500 3962501 3962502 3962520 3962521 3962522 3962600 3962601 3962602 3962620 3962621 3962622
93000-350EA 58270.4 CAD
93000-350 93000-352 93000-354 93000-356 93000-358 93000-360 93000-362 93000-364 93000-366 93000-368 93000-370 93000-372
XPert™ Bulk Powder Filtered Stations, Labconco®
Fume Hoods and Enclosures Balances Enclosures
Bulk powder stations are designed to protect the user from airborne particulates and fumes generated during powder transfer operations
Systems also feature a built-in Guardian™ 1000 digital airflow monitor. If the face velocity falls below the setpoint for more than six seconds, an audible alarm sounds and a red light glows. The alarm can be muted. Airflow monitor features an LCD display of actual face velocity and an RS-232 port. Retaining rings are constructed of Type 304 stainless steel. One retaining ring is required; it should be slightly larger than the diameter of source containerMore Product Information
Order Now
SPECIFICATIONS
Model 5' Wide Enclosures 6' Wide Enclosures
Environmentally Preferable
Exhaust volume 480 cfm (at 100 fpm), 385 cfm (at 80 fpm), 290 cfm (at 60 fpm) 580 cfm (at 100 fpm), 465 cfm (at 80 fpm), 350 cfm (at 60 fpm)
Static pressure loss 0.72–0.85" H2O (at 100 fpm, with HEPA filter), 0.55–0.67" H2O (at 80 fpm, with HEPA filter), 0.38–0.46" H2O (at 60 fpm, with HEPA filter) 0.87–1.03" H2O (at 100 fpm, with HEPA filter), 0.67–0.81" H2O (at 80 fpm, with HEPA filter), 0.46–0.56" H2O (at 60 fpm, with HEPA filter)