XPert™ Bulk Powder Filtered Stations, Labconco®
Supplier: Labconco
3962500 3962501 3962502 3962520 3962521 3962522 3962600 3962601 3962602 3962620 3962621 3962622
93000-350EA 58270.4 CAD
93000-350 93000-352 93000-354 93000-356 93000-358 93000-360 93000-362 93000-364 93000-366 93000-368 93000-370 93000-372
XPert™ Bulk Powder Filtered Stations, Labconco®
Fume Hoods and Enclosures Balances Enclosures
Bulk powder stations are designed to protect the user from airborne particulates and fumes generated during powder transfer operations
Systems also feature a built-in Guardian™ 1000 digital airflow monitor. If the face velocity falls below the setpoint for more than six seconds, an audible alarm sounds and a red light glows. The alarm can be muted. Airflow monitor features an LCD display of actual face velocity and an RS-232 port. Retaining rings are constructed of Type 304 stainless steel. One retaining ring is required; it should be slightly larger than the diameter of source containerMore Product Information