XPert™ Bulk Powder Filtered Systems, Labconco®
Supplier: Labconco
3961500 3961501 3961502 3961520 3961521 3961522 3961600 3961601 3961602 3961620 3961621 3961622
93000-326EA 60126.9 CAD
93000-326 93000-328 93000-330 93000-332 93000-334 93000-336 93000-338 93000-340 93000-342 93000-344 93000-346 93000-348
XPert™ Bulk Powder Filtered Systems, Labconco®
Fume Hoods and Enclosures Balances Enclosures
Bulk powder systems keep airborne particulates and fumes contained during powder transfer procedures
Systems also feature a built-in Guardian™ 1000 digital airflow monitor. If the face velocity falls below the setpoint for more than six seconds, an audible alarm sounds and a red light glows. The alarm can be muted. Airflow monitor features an LCD display of actual face velocity and an RS-232 port. Retaining rings are constructed of Type 304 stainless steel. One retaining ring is required; it should be slightly larger than diameter of source container and a minimum of 5.1cm (2") smallerMore Product Information