CaptairFlow Clean Air Enclosures, Erlab
Supplier: Erlab
GF4.AS GF4.BE GF4.HP FLOW321 FLOW391 FLOW483 FLOW714 PREFILTERXLS
52428-838EA 911.66 CAD
52428-838 52428-840 52428-846 89523-260 89523-262 89523-264 89523-266 10812-246
CaptairFlow Clean Air Enclosures, Erlab
Fume Hoods and Enclosures Laminar Flow Cabinets
Erlab’s CaptairFlow Clean Air Enclosures are designed to prevent cross-contamination for applications such as non-pathogenic cell cultures, in-vitro cultures, microbiology, homeopathic preparations, electronics, and optics where product protection is required. They feature a high efficiency filtration system that provides a particle-free workstation around the manipulation, bringing HEPA filtered clean air down to the work surface.
  • High efficiency UV lights are used to decontaminate the worktop from biological cross-contamination between operations
  • Each
More Product Information