Anti-IRGM Mouse Monoclonal Antibody [clone: 403CT8.5.5]

Supplier: Abgent
AM2003B
89518-742EA 1021.82 CAD
89518-742
Anti-IRGM Mouse Monoclonal Antibody [clone: 403CT8.5.5]
Antibodies
Western Blot: 1:100~500,

Type: Primary
Antigen: IRGM
Clonality: Monoclonal
Clone: 403CT8.5.5
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR