Anti-ESR1 Mouse Monoclonal Antibody [clone: 415CT16.3.2.1]

Supplier: Abgent
AM2006B
89516-700EA 1021.82 CAD
89516-700
Anti-ESR1 Mouse Monoclonal Antibody [clone: 415CT16.3.2.1]
Antibodies
Western Blot: 1:100~250,

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: 415CT16.3.2.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR