Anti-CFC1B Rabbit Polyclonal Antibody

Supplier: Abgent
AP18228A
89516-698EA 1021.31 CAD
89516-698
Anti-CFC1B Rabbit Polyclonal Antibody
Antibodies
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:1000

Type: Primary
Antigen: CFC1B
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol CFC1B
Antigen name cripto, FRL-1, cryptic family 1B
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 653275
Isotype
Western blot Yes
ELISA Yes
Epitope N-Terminal
Immunogen This CFC1B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 20-48 amino acids from the N-terminal region of human CFC1B.
Amino acid number 20 to 48
Sequence REEVTKVATQKHRQC
Molecular weight 24.642 kDa
Purification Protein A & Peptide affinity purification
Concentration 0.5mg/ml
Storage buffer PBS with 0.09% (W/V) sodium azide, pH7.4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR