Anti-ZNF736 Rabbit Polyclonal Antibody

Supplier: Abgent
AP19215A
89515-292EA 1021.31 CAD
89515-292
Anti-ZNF736 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: ZNF736
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ZNF736
Antigen name Zinc Finger Protein 736
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 728927
Isotype
Western blot Yes
ELISA Yes
Epitope N-Terminal
Immunogen This ZNF736 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 49-75 amino acids from the N-terminal region of human ZNF736.
Amino acid number 49 to 75
Sequence KVKRQEAVAKHPC
Molecular weight 49.868 kDa
Purification Protein A & Peptide affinity purification
Concentration 0.5mg/ml
Storage buffer PBS with 0.09% (W/V) sodium azide, pH7.4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR