Anti-GSTT2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP19567C
89514-696EA 1021.31 CAD
89514-696
Anti-GSTT2 Rabbit Polyclonal Antibody
Antibodies
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:1000

Type: Primary
Antigen: GSTT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol GSTT2
Antigen name Glutathione S-transferase Theta 2
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 2953
Isotype
Western blot Yes
ELISA Yes
Epitope Center
Immunogen This GSTT2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 113-141 amino acids from the Central region of human GSTT2.
Amino acid number 113 to 141
Sequence PEEKVERNRTAMDQC
Molecular weight 27.506 kDa
Purification Protein A & Peptide affinity purification
Concentration 0.5mg/ml
Storage buffer PBS with 0.09% (W/V) sodium azide, pH7.4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR