Anti-CHRNB4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20082B
89514-422EA 1021.31 CAD
89514-422
Anti-CHRNB4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CHRNB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR