Anti-ZBTB7A Rabbit Polyclonal Antibody

Supplier: Abgent
AP14208B
89512-736EA 1021.31 CAD
89512-736
Anti-ZBTB7A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50,

Type: Primary
Antigen: ZBTB7A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR