Mini Benchtop Coolers
Supplier: Heathrow Scientific
HS120070 HS120071 HS120072 HS120073 HS120074 HS120075 HS120076
89511-788EA 292.3 CAD
89511-788 89511-790 89511-792 89511-794 89511-796 89511-798 89511-800
Mini Benchtop Coolers
Coolers Benchtop Coolers
Ideal for keeping samples, PCR reagents, enzymes, and other biochemical reagents cool while working at the bench.
  • Protects temperature-sensitive samples at the bench or in the freezer
  • Polycarbonate construction with rubber non-slip feet
  • Gridded and keyed one-position lids for easy sample identification
  • Temperature-retention graph included
Filled with insulating gel, the Mini Coolers can also be used for controlled freezing of samples with longer temperature retention.
Order Now
SPECIFICATIONS
Environmentally Preferable