Avantor® Seradigm Research Grade Fetal Bovine Serum (FBS)
Supplier: VWR
1400-500G 1400-500GH 1400-050 1400-050H 1400-500 1400-500H
97068-070EA 695.09 CAD
97068-070 97068-072 89510-190 89510-192 97068-069 97068-075
Avantor® Seradigm Research Grade Fetal Bovine Serum (FBS)
Sera (Serum)
Avantor® Seradigm Research Grade FBS preserves more of the naturally-occurring growth factors that cells require to thrive, resulting in superior quality sera for a price that fits your budget.
  • Industry-leading collection techniques at the source.
  • Manufacturing techniques reduce bioburden and ensure homogeneity for every bottle and for every lot.
  • Risk mitigation through a robust quality system.
  • Proven track record for performance and consistency.
  • Sourced from countries with health status classified as "Negligible BSE Risk" by the World Health Organization.
More Product Information