PolarSafe™ Stainless Steel Upright Freezer Eco Racks, Argos Technologies
Supplier: Antyila Scientific
04403-50 04403-51 04403-52 04403-53 04403-54 04403-55 04403-56 04403-57 04403-58 04403-59 04403-60 04403-88 04403-89 04403-90 04403-92 04403-93 04403-94 04403-95 04403-61 04403-62 04403-63 04403-64 04403-65 04403-66 04403-67 04403-68 04403-69 04403-70 04403-96 04403-97 04403-98 04403-99 04404-01 04404-03 04404-04 04404-05 04404-06
89508-478EA 204.63 CAD
89508-478 89508-480 89508-482 89508-484 89508-486 89508-488 89508-490 89508-492 89508-494 89508-496 89508-498 89508-500 89508-502 89508-504 89508-506 89508-508 89508-510 89508-512 89508-514 89508-582 89508-584 89508-586 89508-588 89508-590 89508-592 89508-594 89508-596 89508-598 89508-600 89508-602 89508-604 89508-606 89508-608 89508-610 89508-612 89508-614 89508-616 89508-618
PolarSafe™ Stainless Steel Upright Freezer Eco Racks, Argos Technologies
Racks Cryogenic Box Racks
Argos Polarsafe™ Eco Racks are designed to provide an economical alternative to standard freezer racks
The top loading sliding tray racks feature a stainless steel tray versus the more traditional side access drawer racks. Each drawer is segmented to prevent boxes from sticking together as with most tray drawer racks. An easy to grip handle makes it simple to pull out the drawer while wearing protective gloves. Designed for 2" or 3" cardboard or plastic boxes.
The lightweight stainless steel construction allows for easy handling in and out of a freezer. Racks will accommodate cardboardMore Product Information