Gastight® CTC PAL® S-Line Syringes for HPLC Autosamplers, Hamilton
Supplier: Hamilton
67444-01 67446-01 67450-01 67452-01 67442-01 67448-01
89498-972EA 190.67 CAD
89498-972 89498-974 89498-976 89498-978 89498-980 89498-982
Gastight® CTC PAL® S-Line Syringes for HPLC Autosamplers, Hamilton
Syringes Syringes for Chromatography
CTC PAL autosamplers are one of the most popular sampling devices on the market today
S-Line syringes are the cost-effective choice for CTC PAL autosampler applications. S-Line syringes deliver great performance in everyday use for HPLC injections at an exceptional value. The Gastight® S-Line syringe is an ideal choice for HPLC sample injections. It incorporates a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit in the glass barrel, the tip wipes the interior of the syringe barrel free of sample, minimizing sample carryover. This feature isMore Product Information