Samco™ Bio-Tite™ Specimen Container, 60 ml/48 mm (2 oz.), Thermo Scientific
Supplier: Samco
03 0006 03 0007 03 0193 03 0195
89498-364CS 661.56 CAD
89498-364 89498-366 89498-380 89498-382
Samco™ Bio-Tite™ Specimen Container, 60 ml/48 mm (2 oz.), Thermo Scientific
Sample Containers General Purpose Containers
Prevent bio-hazardous spills with the leak-resistant Samco™ Bio-Tite™ Wide-Mouth 60mL (2oz.) 48mm Specimen Containers
  • Plug seal cap ensures excellent leak-resiatant performance
  • Cap and base knurls ensure ease of handling
  • Available either assembled or in bulk packed format ideal for pre-fill applications
  • Supplied nonsterile or gamma irradiated for guaranteed sterility
These stackable, shatter-resistant containers are manufactured from virgin polypropylene and are ideal for the collection and transportation of both liquid and solid samples. Available inMore Product Information