VWR® Extreme Temperature Tape
Supplier: VWR International
89495-448 89495-450 89495-452
89495-448CS 2800.57 CAD
89495-448 89495-450 89495-452
VWR® Extreme Temperature Tape
Tapes
This Extreme Temperature Tape is a transparent amber polyimide film with pressure sensitive silicone adhesive.
  • Transparent amber polyimide film
  • Wound on a plastic core
The extreme temperature-resistant properties (-100°F to 500°F) of this tape make it suitable for high heat and sub zero applications
Recommended applications include flame-resistent electrical insulation, solder mask for circuit boards, heat sealing, lamination, cold storage, lyophilizing, freeze drying, and over-labeling.
The silicone adhesive ensures a clean release from mostMore Product Information