VWR® Sterile Bucket Liners
Supplier: VWR International
89495-436 89495-438 76304-356
89495-436CS 468.49 CAD
89495-436 89495-438 76304-356
VWR® Sterile Bucket Liners
Bags Waste Disposal Bags
These LDPE/nylon blend bucket liners/bags create an impermeable barrier between strong disinfectant solutions and buckets.
  • Sterile with sterility level of assurance (SAL 10⁻⁶)
  • LDPE/nylon blend fabrication
  • Provide an impermeable barrier against strong disinfectant solutions
This additional step ensures the integrity of the disinfectant blend. Item 89495-436 can be used for transport of sterile equipment.
Order Now
SPECIFICATIONS
Environmentally Preferable