Anti-GST Tag Mouse Monoclonal Antibody [clone: 2F10B9] (HRP (Horseradish Peroxidase))
Supplier: Genscript
A00866-100
89493-656EA 314.83 CAD
89493-656
Anti-GST Tag Mouse Monoclonal Antibody [clone: 2F10B9] (HRP (Horseradish Peroxidase))
Antibodies
THE™ GST antibody [HRP], mAb, Mouse recognizes C-terminal, N-terminal and internal tagged GST fusion proteins.
  • Comprehensive pool
  • Against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufacturedMore Product Information