Buchner Funnels, Porcelain, Chemglass
Supplier: Chemglass
CG-1888-01 CG-1888-02 CG-1888-03 CG-1888-04 CG-1888-05 CG-1888-06 CG-1888-07 CG-1888-08 CG-1888-09
89492-796EA 103.06 CAD
89492-796 89492-798 89492-800 89492-802 89492-804 89492-806 89492-808 89492-810 89492-812
Buchner Funnels, Porcelain, Chemglass
Funnels Filtration Funnels Büchner Funnels
Buchner funnel with fixed perforated plate, glazed both inside and outside with the exception of the top rim
  • Follow gradual heating/cooling rates
  • Avoid contact of heated ware with a cold surface
  • Use an oven or hot plate and an intermediate step when quicker heating/cooling rates are required
  • Carefully inspect labware prior to each use and do not use any product that appears defective
The Porcelain glassware is widely used in the chemistry laboratory. It has excellent resistance to all reagents except hydrofluoric acid, and is highly resistant toMore Product Information