Titrette® Bottle Top Burets, Chemglass
Supplier: Chemglass
CG-1161-100 CG-1161-200
89492-746EA 3289.85 CAD
89492-746 89492-748
Titrette® Bottle Top Burets, Chemglass
Burettes
The CG-1161 series Titrette® is the new generation of bottle top burette from BRAND
Features include Class A precision, Smooth, Low-force operation, low-profile structure, and chemical compatibility.
It has a number of innovative features and upgrades for more accurate and convenient titration. The Titrette® avoids the risk of spills from poured transfers to glass or plastic burets. The instrument is well suited for general chemistry, water treatment applications, food and beverage analysis, industrial titrations and environmental work.
Ordering information: Complete with a bottleMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable