Vacuum Round Bottom Flask with Hi-Vac Valve Sidearm, Ace Glass Incorporated
Supplier: Ace Glass
7764-02 7764-04 7764-06 7764-08 7764-10
89428-086EA 364.46 CAD
89428-086 89428-088 89428-090 89428-092 89428-094
Vacuum Round Bottom Flask with Hi-Vac Valve Sidearm, Ace Glass Incorporated
Flasks Nitrogen Flasks
Round bottom borosilicate glass flask with standard taper center neck and a 0-4 Hi-Vac valve for precise control
Flask is ideal for simple NO-AIR and vacuum applications. Outlet arm is 8 mm and can be fitted with a rubber septa for insertion of samples or additives via syringe.
Order Now
SPECIFICATIONS
Environmentally Preferable