Low Vacuum Stirrer Bearings, Ace Glass Incorporated
Supplier: Ace Glass
8050-02 8050-04 8050-06 8050-08 8050-10 8050-12 8050-14 8050-16 8050-18
71001-558EA 1918.59 CAD
71001-558 71001-560 71001-562 71001-564 71001-566 71001-568 89427-874 89427-876 89427-878
Low Vacuum Stirrer Bearings, Ace Glass Incorporated
Stirrers Stirring Shafts
Low Vacuum Stirrer Bearings are designed for use in applications to below 3 Torr.
All PTFE, Rulon, or PEEK wetted area construction provides for low extractables. Completely contained bearing and internal parts also provide non-shedding of materials into the reaction. The bearing is excellent for vacuum applications and can be operated at sub-torr levels. Rated for up to 400rpms. It can be used on either glass polished or stainless steel stir shafts.
Ordering information: Stirrer Bearings 71001-558, 71001-560, 71001-566, and 71001-568 utilize replacement FETFE O-Ring 89061-002 (size 116More Product Information