Anti-IgG Chicken Polyclonal Antibody (CF™ 568)

Supplier: Biotium

20337-50UL
89427-180EA 137.18 CAD
89427-180
Anti-IgG Chicken Polyclonal Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 568
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgY
Reactivity: Goat
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antigen name Immunoglobulin G
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 568
Reactivity Goat
Host Chicken
Isotype IgY
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Epitope Heavy and Light chain

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR