Recovery Flask, Glassco
Supplier: Glassco
069.470.22
89426-820EA 110.92 CAD
89426-820
Recovery Flask, Glassco
Flasks Round Bottom Flasks
Manufactured from Borosilicate 3.3 glass
All products listed are hand blown from tubing to ensure uniform thickness.