PFA Bottles, Savillex
Supplier: SAVILLEX
150-01-0100 150-01-0250 150-01-0500 150-01-1000 150-01-0050
30617-482EA 459.04 CAD
30617-482 30617-484 30617-486 30617-488 89422-786
PFA Bottles, Savillex
Bottles Plastic Bottles
PFA bottles are made with virgin, high-purity Perfluoroalkoxy (PFA) resin and manufactured by a proprietary stretch blow molding process, adapted for fluoropolymer use by Savillex.
  • Non-Cytotoxic
  • Extractables report available from Savillex
The accessory PFA GL45 Transfer Closure is designed for use with PFA or FEP bottles sized 100 to 2000 ml. The closure features two ¼" tube ports (one top and one side) and includes ¼" ferrule nuts. It can be used to connect bottles to the Savillex Acid Purification System, or for other liquid transfer applications.
More Product Information