TGL900 Series Class 100 Cleanroom Latex Glove, TechNiGlove
Supplier: Techniglove
TGL901 TGL902 TGL903 TGL904
89415-098CS 658.64 CAD
89415-098 89415-100 89415-102 89415-104
TGL900 Series Class 100 Cleanroom Latex Glove, TechNiGlove
Gloves
Techni-Cleaned ambidextrous 9.5" Class 100 powder free latex gloves are designed for controlled environments requiring stringent low particles and extractables.
  • Class 100
  • 9.5" ambidextrous
  • Powder-free
Techniglove products are made from th highest grade latex available. They contain no silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive On-line Batch Testing is available for this product.
Certifications: COC available onMore Product Information