TechNiCleaned ESD Series Class 100 Nitrile Gloves, TechNiGlove
Supplier: Techniglove
TN2000W-E TN2001W-E TN2002W-E TN2003W-E TN2004W-E TN2005W-E
89415-006CS 882.45 CAD
89415-006 89415-008 89415-010 89415-012 89415-014 89415-016
TechNiCleaned ESD Series Class 100 Nitrile Gloves, TechNiGlove
Gloves
TechniCleaned Ambidextrous 12" Class 100 powder-free nitrile gloves are designed for controlled environments requiring stringent low particles and extractables.
  • Proprietary ESD formulation
  • Class 100
  • 100% Nitrile
  • 12" ambidextrous
  • Powder-free
  • For us in semiconductor, disc drive, flat panel, electronic, and pharmaceutical manufacturing
Techniglove products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel.More Product Information