HBIO AgilePulse In Vivo Electroporation System (ID and IM), BTX

Supplier: Harvard Apparatus
47-0400N 47-0401N 47-0500N 47-0501N 47-0200N 47-0201N
89414-554EA 43936.29 CAD
89414-554 89414-556 89414-558 89414-560 89414-562 89414-564
HBIO AgilePulse In Vivo Electroporation System (ID and IM), BTX
Electroporation Systems Electroporators
Providing maximum efficiency DNA and RNA delivery to skin, muscle, and tumor tissues. Used for animal immunization, vaccine development and gene therapy, provides an intra-muscular, intra-dermal or intra-tumor electroporation solution to produce maximum transfection efficiency.

  • Simple user interface
  • Uniform, reliable electroporation
  • Simple, fast treatment
  • Optimal design

The AgilePulse In Vivo System, used for animal immunization, vaccine development, and gene therapy, provides an intra-muscular, intra-dermal, or intra-tumor electroporationMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Interfaces USB flash key
Voltage range 50 - 1200 V
Pulse width 0.050 - 10 ms
Pulse interval 0.200 - 1000 ms (5 kHz - 1 Hz)
Display Touch Screen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR