Insulated Shippers, Expanded Polystyrene (EPS), Foam Only and with Corrugate
Supplier: Sonoco Thermosafe
315UPS 317UPS 346UPS 446UPS 447UPS 448UPS 609UPS 493 498 614 617UPS 613 656 615 453 494 438 439 326 806 397 479 499 411 452 425 430 413 333 676 648 684 68-9LB4 79-8LB4 1714-12LB4 56-8LB4 1212-10LB4 68-4LB4 647 802 812 322UPS 325UPS 321UPS 324UPS 348UPS 608UPS 457 482 316 318 319 355 398 352 121212VWR 675 637 638 831 832 669 803 804 481 431UPS 433 801 809 351 655 634 635UPS 657 658 663 682 683 477 612 610 611 661 662 489UPS 490UPS 685 653 654 651 649 622UPS 618UPS 619UPS 616UPS 680 681 646 428 68-12LB4 68-9NB4 1017-11LB4 175LB4 110LB4 1714-12NB4
14100-406CS 374.22 CAD
14100-406 14100-410 14100-418 14100-428 14100-430 14100-432 14100-434 14100-448 14100-452 14100-454 14100-458 14100-474 14100-482 15713-510 15713-543 15713-551 15713-607 15713-608 15713-623 15713-664 15713-672 15714-465 15714-468 15714-486 15714-506 15714-511 15714-513 15714-522 73320-188 73320-200 82028-388 82028-390 89050-250 89050-252 89050-254 89050-256 89050-258 89050-260 95042-406 95054-978 95054-980 14100-400 14100-402 14100-412 14100-414 14100-420 14100-436 15713-544 15713-545 15713-600 15713-700 15713-702 15714-500 15714-704 89179-608 89260-322 89370-076 89413-338 89413-340 89413-342 89413-348 89413-350 89413-362 89413-370 89413-372 89413-374 89413-378 89413-380 89413-384 89413-386 89413-394 89413-396 89413-398 89413-400 89413-402 89413-404 89413-414 89413-422 89413-424 89413-432 89413-434 89413-436 89413-438 89413-450 89413-452 89413-454 89413-456 89413-464 89413-466 89413-468 89413-470 89413-472 89413-476 89413-478 89413-480 89413-482 89413-486 89413-488 89413-490 89495-994 10770-536 10770-538 10770-596 10770-598 75817-446 76006-454 76006-455
Insulated Shippers, Expanded Polystyrene (EPS), Foam Only and with Corrugate
Mailers and Shippers
ThermoSafe's extensive line of EPS foam-only and EPS foam with corrugate insulated shippers offers the ideal solution for the safe storage and delivery of most refrigerated or frozen shipping products.
  • Large selection available
  • Excellent dry ice sublimation
A large selection of cost-effective, reusable EPS foam-only and EPS foam in corrugate stock containers are available. Excellent dry ice sublimation; average dry ice usage in 1½" thick wall units is 5 lbs. over 24 hours and 3 to 4 lbs. thereafter. Standard 1½" wall shippers are ideal for shipmentsMore Product Information