Tygon® Ultra Soft Tubing, Formulation E-1000, Saint-Gobain Life Sciences
Supplier: Saint Gobain Life Sciences
ADK00027 ADK00038
89412-462EA 102.4 CAD
89412-462 89412-464 89412-466 89412-468 89412-470 89412-472 89412-474 89412-478 89412-480
Tygon® Ultra Soft Tubing, Formulation E-1000, Saint-Gobain Life Sciences
Tubing Transfer Tubing
Tygon® E-1000 is a non-DEHP laboratory tubing that meets the market demand for safety and quality.
The sterilization method for this tubing is ethylene oxide gas.
Tygon® E-1000 tubing is soft and flexible for lab set-ups. It offers low-temperature resistance for low level cryogenic applications. It is also resistant to corrosive chemicals for laboratory applications.
This tubing is an excellent solution to applications requiring complicated laboratory set-ups with sharp radius curves and multiple direction changes. Tygon® E-1000 can resist twisting, collapsing, and remains flexibleMore Product Information
Order Now
SPECIFICATIONS
Model 89412-462 89412-464 89412-466 89412-468 89412-470 89412-472 89412-474 89412-478 89412-480
Durometer hardness Shore A, 40 Shore A, 40 Shore A, 40 Shore A, 40 Shore A, 40 Shore A, 40 Shore A, 40 Shore A, 40 Shore A, 40
Environmentally Preferable
Max. vacuum 29.9 29.9 15.0 7.0 29.9 5.0 3.0 2.0 7.0
Working pressure 26.0 psi 16.0 psi 10.0 psi 8.0 psi 14.0 psi 7.0 psi 7.0 psi 6.0 psi 8.0 psi