Tweezers, Straight Fine Tips, Style 3, Excelta Corp®
Supplier: Excelta
3-S 3-SA-PI 3-SA-SE 3-S-PI 3-S-SE 64-SA 3-SA-PI-ET 3-SA-ET
63042-142EA 127.43 CAD
63042-142 63042-144 63042-148 63042-150 63042-152 63042-154 63042-718 89411-770 89411-824
Tweezers, Straight Fine Tips, Style 3, Excelta Corp®
Forceps
Style 3 is ideal for precision work, including under magnification.
  • Ideal for working under magnification
  • Available in three different materials
  • Available in three different qualtiy grades
  • Ergonomic handles are standard option
  • Teflon coated tip option for non-marring
The 'Style 3' series straight very fine precision pointed tweezer is designed for work under magnification. The overall length is 4.75". These are also availabe with ergonomic handles (ET). We offer 3 different qualities. The higher the quality, the more precise the tips. They are offeredMore Product Information