Tweezers, Straight Strong Tips, Style 00, Excelta Corp®
Supplier: Excelta
000-SA 000-SA-PI 00B-SA 00B-SA-SE 00C 00C-SA 00C-SA-SE 00D 00D-SA-PI 00D-SA-SE 00-PI 00-SA-ET 00-SA-PI 00-SA-SE 00-SE 0A 0C-SA-PI 0C-SA-SE 0-SA 0-SA-SE 00B-SA-PI 00-SA-PI-ET 00D-SA-PI-ET 00-S 00D-SA-ET 00-SA-R 00D-SA-SE-ET
63041-006EA 145.03 CAD
63041-006 63041-008 63041-012 63041-014 63041-016 63041-018 63041-020 63041-024 63041-028 63041-030 63041-032 63041-036 63041-038 63041-040 63041-044 63041-046 63041-048 63041-050 63041-052 63041-062 63041-064 63041-066 89411-698 89411-760 89411-766 89411-794 89411-812 89411-828 76471-446 76471-448 76471-450
Tweezers, Straight Strong Tips, Style 00, Excelta Corp®
Forceps
This thick and medium pointed tweezer is excellent for general assembly.
  • Strong tips, won't bend
  • Most popular general assembly tweezer
The 'Style 00' Series straight medium pointed tweezers is the most popular general purpose tweezer used without magnification. The series features robust medium pointed tips for longevity and a variety of choices, like ergonomic handles, serrated tips, radius edges and fluted handles, depending on your application. Three different qualities are offered. The higher the quality, the more precise the tips. They areMore Product Information