ULTRA-TURRAX® T10 Basic Disperser, IKA® Works
Supplier: IKA Works
3737000
89409-872EA 4410.24 CAD
89409-872 89409-874
ULTRA-TURRAX® T10 Basic Disperser, IKA® Works
Homogenizers Benchtop Homogenizers
Small dispersing instrument designed for use with volumes ranging from 0.5 to 100mL (H₂O)
Units are equipped with quick-connect couplings for changing dispersing elements easily. Ideal for manual operation due to its lightweight and ergonomic design, the T10 Basic Disperser is also extremely mobile due to direct line power (no transformer required). Stainless steel dispersing elements can be cleaned quickly and easily, and they can be dismounted without tools. Plastic disposable dispersing elements are available in two sizes, particularly suitable for PCR analysis. High performance driveMore Product Information