TRACK-IT™ Relative Humidity and Temperature Logger, Monarch Instrument
Supplier: Monarch Instrument
5396-0201 5396-0202 5396-0203 5396-0204 5396-0201-CAL 5396-0202-CAL 5396-0203-CAL 5396-0204-CAL
89184-574EA 308.64 CAD
89184-574 89184-576 89184-578 89184-580 89409-784 89409-786 89409-788 89409-790
TRACK-IT™ Relative Humidity and Temperature Logger, Monarch Instrument
Data Loggers
The RH/Temp data logger is a battery powered stand alone compact data logger that records up to 32,000 samples each of relative humidity and temperature data.
  • Push button or program to start recording
  • Alarm set points and one year battery life
  • N.I.S.T. calibration available
  • High accuracy, small compact size and repeatability
  • Accurate internal clock (one minute/year)
  • Dew point calculation standard
  • Multiple user programmable display
The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. The on board data storage isMore Product Information