Tygon S3™ B-44-3 Beverage Tubing, Saint-Gobain Life Sciences
Supplier: Saint Gobain Life Sciences
AAB00027 AAB00069 AAB00003 AAB00005 AAB00007 AAB00009 AAB00011 AAB00012 AAB00013 AAB00016 AAB00018 AAB00019 AAB00022 AAB00023 AAB00024 AAB00028 AAB00029 AAB00036 AAB00038 AAB00046 AAB00053 AAB00062 AAB02002 AAB02007 AAB04002
63010-960PK 112.42 CAD
63010-960 89404-086 89404-102 89404-108 89404-122 89404-130 89404-132 89404-134 89404-136 89404-138 89404-140 89404-152 89404-064 89404-066 89404-070 89404-072 89404-074 89404-076 89404-078 89404-080 89404-082 89404-084 89404-088 89404-090 89404-092 89404-094 89404-096 89404-098 89404-100 89404-104 89404-106 89404-110 89404-112 89404-114 89404-116 89404-118 89404-120 89404-124 89404-126 89404-128 89404-142 89404-144 89404-148 89404-154
Tygon S3™ B-44-3 Beverage Tubing, Saint-Gobain Life Sciences
Tubing Transfer Tubing
Tygon S3™ B-44-3 Beverage Tubing from Saint-Gobain Performance Plastics is now phthalate-free.
Tygon S3™ B-44-3 Beverage Tubing is as clear as glass for easy visual monitoring of flow. Lightweight and flexible for easy and quick installation. Broad chemical resistance. Non-wetting properties allow easy cleaning and complete drainage.
Saint-Gobain is proud to be among the first companies to offer sustainable flexible tubing products. This bio-based tubing line combines the high performance standards customers demand with an eco-friendly design. Versatile, high-performance Tygon S3™ B-44-More Product Information