VWR® Inverted Fluorescence Microscope
Supplier: VWR International
89403-564 89403-642 89403-648 89403-652 89403-656 89403-658 89403-662 89403-664 89404-464
89403-564EA 148.8 CAD
89403-564 89403-642 89403-648 89403-652 89403-656 89403-658 89403-662 89403-664 89404-464
VWR® Inverted Fluorescence Microscope
Microscopes
The VWR® Inverted Fluorescence Microscope is ideal for applications in biology, cell cytology, genetics, oncology, and immunology, and it is also well suited for research, universities, routine laboratory, and medical demands.
  • Ideal for biology, cell cytology, genetics, oncology, and immunology applications
  • Trinocular Siedentopf tube with 45° inclination
  • One-year warranty on electrical parts
  • Five-year warranty on mechanical parts and optics
The VWR® Inverted Fluorescence Microscope features Siedentopf trinocular tube with a 45° inclination (LightMore Product Information