EVAHL PAPRs (Powered Air Purifying Respirators), Blower Assemblies, Bullard®
Supplier: Bullard
EVAHL1 EVAHL2 EVAHL3 EVAHL4
89402-894EA 1266.7 CAD
89402-894 89402-896 89402-898 89402-900
EVAHL PAPRs (Powered Air Purifying Respirators), Blower Assemblies, Bullard®
Respirators Respiratory Systems
The EVAHL is simple, safe, and dependable. The system features one-button operation, a choice of two air speeds, and a single filter design.
  • Certified for hazardous locations (Class I Division 2, Class II Division 2, Class III)
  • Versatile respirator for particulate or chemical environments
  • Lightweight, compact, well-balanced design
Additionally, the EVAHL offers Active Flow Technology™, which maintains an airflow of 8.5 CFM (high speed), 1000 Assigned Protection Factor (depending upon head top), and dual alarms for low flow and low battery. Finally,More Product Information