Zirconium Crucibles
Supplier: XRF Scientific
ZXS25 ZXS30 ZXS35 ZXS45 ZXS50 ZXS75 ZXS100
89401-344EA 273.31 CAD
89401-344 89401-346 89401-348 89401-352 89401-354 89401-356 89401-360
Zirconium Crucibles
Crucibles
Zirconium's distinctive properties make it a popular choice for fusions and fluxes in analytical chemistry and chemical processing.
These standard shape Zirconium crucibles are highly resistant to corrosion. They do not require handling with platinum tipped tongs and they can be cleaned with most common agents (except hydrofluoric acid). Zirconium crucibles suffer little to no attack or oxidation and are resistant to melts of alkali carbonates, hydroxides, peroxides, borates, nitrate chlorides, and some fluorides. This construction virtually eliminates the problem of sample contamination.More Product Information