Platinum Dishes; Standard and Evaporation Shapes, XRF Scientific
Supplier: XRF Scientific
PT99GD50L PT99GD75L PT99GD100L PT99GD125L PT99GD150L PT99GD200L PT99GD250L PT99GD50LLID PT99GD75LLID PT99GD100LID PT99GD125LID PT99GD150LID PT99GD200LID PT99GD250LID PT99GED100 PT99GED125 PT99GED150
89401-304EA 2418.65 CAD
89401-304 89401-306 89401-308 89401-310 89401-312 89401-314 89401-316 89401-318 89401-320 89401-322 89401-324 89401-326 89401-328 89401-330 89401-332 89401-334 89401-336
Platinum Dishes; Standard and Evaporation Shapes, XRF Scientific
Trays/Dishes/Pots Evaporating Basins/Dishes
These 99.95% pure platinum dishes are available in standard and evaporation forms.
  • High temperature strength
  • Ductility; corrosion; and oxidation resistance
  • Excellent resistance to chemical attacks by acids and fusion mixtures
Due to their high temperature strength; ductility; corrosion and oxidation resistance; platinum dishes are critical when high temperatures and resistance to chemical attacks are required.
Ideally suited for fusions; wet chemistry; digestion; ash testing; and Loss on Ignition (LOI) applications.
Certifications: The 100 mlMore Product Information