Platinum Crucibles; Standard and Wide Shapes, XRF Scientific
Supplier: XRF Scientific
PT99GCS05L PT99GCS10L PT99GCS15L PT99GCS20L PT99GCS25L PT99GCS30L PT99GCS35L PT99GCS40L PT99GCS50L PT99GCS100L PT99GCS150L PT99GCS200L PT99GCS250L PT99GCS05LLID PT99GCS10LLID PT99GCS15LLID PT99GCS20LLID PT99GCS25LLID PT99GCS30LLID PT99GCS35LLID PT99GCS40LLID PT99GCS50LLID PT99GCS100LID PT99GCS150LID PT99GCS200LID PT99GCS250LID PT99GCLC10L PT99GCLC15L PT99GCLC20L PT99GCLC25L PT99GCLC30L PT99GCLC35L PT99GCLC40L PT99GCLC50L PT99GCLC100L PT99GCLC10LID PT99GCLC15LID PT99GCLC20LID PT99GCLC25LID PT99GCLC30LID PT99GCLC35LID PT99GCLC40LID PT99GCLC50LID PT99GCL100LID
89401-216EA 1504.9 CAD
89401-216 89401-218 89401-220 89401-222 89401-224 89401-226 89401-228 89401-230 89401-232 89401-234 89401-236 89401-238 89401-240 89401-242 89401-244 89401-246 89401-248 89401-250 89401-252 89401-254 89401-256 89401-258 89401-260 89401-262 89401-264 89401-266 89401-268 89401-270 89401-272 89401-274 89401-276 89401-278 89401-280 89401-282 89401-284 89401-286 89401-288 89401-290 89401-292 89401-294 89401-296 89401-298 89401-300 89401-302
Platinum Crucibles; Standard and Wide Shapes, XRF Scientific
Crucibles
These 99.95% pure platinum crucibles are available in standard and wide forms.
Due to their high temperature strength; ductility; corrosion and oxidation resistance; platinum crucibles are critical when high temperatures and resistance to chemical attacks are required.
High temperature strength
Ductility; corrosion; and oxidation resistance
Excellent resistance to chemical attacks by acids and fusion mixtures
Ideally suited for fusions; wet chemistry; digestion; ash testing; and Loss on Ignition (LOI) applications.
Certifications: The 15; 20; 25; and 30mL Standard Shape cruciblesMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable