Anti-LHX3 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX14555
89360-036EA 1052.09 CAD
89360-036
Anti-LHX3 Rabbit Polyclonal Antibody
Antibodies
The pituitary gland releases hormones that regulate growth, lactation, homeostasis, the stress response, reproductive development and fitness as well as water metabolism. LHX3 is one of several transcription factors that have been implicated in pituitary development. This particular transcription factor is critical for motor neuron specification and the development of the anterior and intermediate lobes of the pituitary gland in mammals. The sequence of the human LHX3 gene has been identified and characterized by IU researchers.

ICC: Use at a dilution of 1/100 - 1/250. IF: Use at an assayMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol LHX3
Environmentally Preferable
Antigen name LIM Homeobox 3
Antigen synonyms DKFZp762A2013|CPHD3|LIM3|M2-LHX3
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Fish
Host Rabbit
Gene ID 16871
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Immunogen C-terminal portion of mouse LHX3 protein.
Purification Protein A purified
Concentration 0.5 mg/ml
Storage buffer Preservative: 0.1% Sodium Azide; Constituents: PBS. pH 7.2
Storage temperature Add glycerol to a final volume of 50%, aliquot and store at –20 °C. Avoid repeated freeze / thaw cycles.
Tested applications ICC/IF, IHC-P, IHC-Fr, IP, WB

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR