Anti-SOD1 Rabbit Polyclonal Antibody
Supplier: Genetex
GTX13499
89359-696EA 995.32 CAD
89359-696
Anti-SOD1 Rabbit Polyclonal Antibody
Antibodies
Superoxide dismutase (SOD) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells. They are: extracellular SOD (EC SOD), copper and zinc-containing SOD (CuZn SOD) and manganese-containing SOD (Mn SOD). The CuZn form contains Cu and Zn ions and exists as a 32 kDa dimer in the cytosol. Mn SOD is an 80 kDa tetramer that contains Mn ion and resides in the mitochondrial matrix. Mn SOD is a tumor necrosis factor (TNF)- inducible enzyme that protectsMore Product Information