Anti-SSTR2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX13120
89359-596EA 994.15 CAD
89359-596
Anti-SSTR2 Rabbit Polyclonal Antibody
Antibodies
Somatostatin is a tetradecapeptide that is widely distributed in the body. It acts on multiple organs in the body and also functions as a neuropeptide affecting electrical activity of neurons. Somatostatin Receptor 2 (SSR2), along with SSR1, is expressed at the highest levels in the stomach and jejunum, cerebrum and kidney, respectively.

IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SSTR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR