Anti-MED17 Rabbit Polyclonal Antibody
Supplier: Genetex
GTX115241
89358-292EA 939.18 CAD
89358-292
Anti-MED17 Rabbit Polyclonal Antibody
Antibodies
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TRMore Product Information