Anti-SF2 Rabbit Polyclonal Antibody
Supplier: Genetex
GTX114918
89358-184EA 939.18 CAD
89358-184
Anti-SF2 Rabbit Polyclonal Antibody
Antibodies
This gene encodes a member of the arginine/serine-rich splicing factor protein family, and functions in both constitutive and alternative pre-mRNA splicing. The protein binds to pre-mRNA transcripts and components of the spliceosome, and can either activate or repress splicing depending on the location of the pre-mRNA binding site. The protein's ability to activate splicing is regulated by phosphorylation and interactions with other splicing factor associated proteins. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]