Anti-ATP6V1E1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX114056
89357-856EA 939.18 CAD
89357-856
Anti-ATP6V1E1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ATP6V1E1 (ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ATP6V1E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR