Anti-PTPRR Rabbit Polyclonal Antibody

Supplier: Genetex
GTX111152
89356-594EA 939.18 CAD
89356-594
Anti-PTPRR Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PCPTP1 (protein tyrosine phosphatase, receptor type, R)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PTPRR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR